Anti-EGFR Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: AY13]

Supplier: Biolegend
352909 352910
10759-650EA 199.1 USD
10759-650 10759-652
Anti-EGFR Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: AY13]
Antibodies
PE/Cy7 anti-human EGFR [AY13]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: AY13
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR