Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11]

Supplier: Biolegend
355201 355202
10759-594EA 139.6 USD
10759-594 10759-596
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11]
Antibodies
Purified anti-human SSEA-5 [8E11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC, IHC, IP; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSEA-5
Clonality: Monoclonal
Clone: 8E11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR