QBC® STAR™ Dry Hematology Analyzer, Drucker Diagnostics
Supplier: Drucker
429001
10755-958EA 23590.42 USD
10755-958
QBC® STAR™ Dry Hematology Analyzer, Drucker Diagnostics
Clinical Diagnostic Supplies Clinical Analysers
The QBC STAR™ dry hematology system is the gold standard for performing CBC testing in a variety of settings, including office labs and even remote locations. It employs innovative dry hematology technology, meaning that all of the reagents needed to perform a CBC test with it are contained in the QBC STAR™ tube. After filling the tube with the blood samples, the clinician simply places it in the specially designed QBC STAR™ system and obtains the test results after the analysis is complete. This unique approach provides multiple benefits for in-office CBC testing.
  • Fully automatic
  • Simple, single button operation
  • No liquid reagents
  • Nine parameter, two part differential CBC
  • For capillary and venous blood
  • Accurate reads without the calibration
  • Built-in printer prints results on 58 mm thermal recorder paper
  • 3 external USB ports
The testing items include hematocrit (Hct) (%), hemoglobin (Hgb) (g/dl).
One-year warranty