Proteinase K
Supplier: Analytik Jena US
845-CH-0010030
10755-738EA 67.72 USD
10755-738
Proteinase K
Enzymes
innuPREP proteinase K is a highly active recombinant protease from Pichia pastoris with endo-and exoproteolytic activity.
  • Contains no RNases or DNases and virtually no DNA
  • Consistent quality and performance
  • Stable over a broad pH range from 4 to 12.5
  • Ideal for preparing cell lysates for subsequent nucleic acid isolation
This enzyme is extraordinarily effective against native proteins and can be used for quickly inactivating endogenous RNases and DNases. Proteinase K is suitable for isolating nucleic acids for use in amplification reactions, for isolating native RNA and DNA form tissues and cell lines, for promoting cell lysis by activating a bacterial autolysis factor and for modifying proteins and/or glycoproteins on cell surfaces (for membrane structure analyses).