VACUETTE® Low Volume Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
454081
10755-336CS 219 USD
10755-336 10755-700 10755-702
VACUETTE® Low Volume Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Greiner Bio-One offers a complete line of low volume tubes with draw volumes of 1 to 2.5 mL to facilitate reduction of phlebotomy volumes, improve overall patient care, and minimize the amount of discarded specimen waste.
  • Minimizes discarded specimen waste
  • Low level of volume required for samples
  • Color-coded safety caps
All low volume tubes are standard 13x75mm plastic tubes with color-coded safety caps. Tubes offer functional compatability with instrumentation.