BAX® System Protease, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
ASY2012
10754-938EA 37.05 USD
10754-938
BAX® System Protease, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
Fast and accurate molecular method for pathogen detection and more.
  • Advanced, automated testing
  • Breaks down samples ar rhe genetic level
  • Load it and leave it operation
Fast and accurate molecular method for pathogen detection and more
The award-winning BAX® System breaks down samples at the genetic level, using the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes with certainty. These highly reliable results help food companies make product release decisions with confidence.
Customers around the world have made the BAX® System an integral part of their quality control systems due to its tremendous impact on their operations--from dramatically decreasing false positives and minimizing re-testing, to reducing employee training and speeding the time to market.
The speed and accuracy of DNA-based detection, including a license to perform PCR. The simplicity of an automated load-it and leave-it operation. The familiarity of a Microsoft® Windows® interface. The BAX® System combines it all for smooth testing procedures at any stage in the processing pipeline.