BAX® System Real-Time PCR Assay for Vibrio cholerae / parahaemolyticus / vulnificus, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
KIT2010
10754-892EA 1567.97 USD
10754-892
BAX® System Real-Time PCR Assay for Vibrio cholerae / parahaemolyticus / vulnificus, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The Hygiena™ BAX® System PCR Assay for Vibrio cholerae, parahaemolyticus, and vulnificusproduces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
The Hygiena BAX System Real-Time PCR Assay for Vibrio cholerae, parahaemolyticus, and vulnificus produces clear yes-or-no results for three major Vibrio strains in less than 24 hours after just 70 minutes of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for presence/absence or MPN testing for tuna, raw and cooked shrimp, scallops, and oysters.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.