BAX® System Real-Time PCR Assay for Shigella, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
KIT2007
10754-886EA 1742.2 USD
10754-886
BAX® System Real-Time PCR Assay for Shigella, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
The Hygiena™ BAX® System PCR Assay for Shigella produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
The Hygiena BAX System Real-Time PCR Assay for Shigella is a screening assay designed to quickly eliminate negative results, reducing the number of samples that must be confirmed. This assay produces clear yes-or-no results after about 65 minutes of processing in the BAX System Q7 instrument. The BAX System Real-Time PCR Assay for Shigella offers fewer presumptive positive results than traditional methods, which means reduced hands-on time and cost for culture confirmations.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.