BAX® System PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Supplier: Hygiena
KIT2000 KIT2004
10754-878EA 1795.95 USD
10754-878 10754-898
BAX® System PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Microbiology Test Systems PCR Tests
TheHygiena™ BAX® System PCR Assay for E
Delivering results in as few as 10 hours for some matrices, the DuPont BAX System Real-Time PCR Assay for E. coli O157:H7 produces clear yes-or-no results for all known E. coli O157:H7 -including rough strains - after about 65 minutes of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for raw meat and produce samples. Validated by AFNOR Certification for raw beef and raw vegetable samples. Adopted by the USDA-FSIS for meat products, carcass, and environmental sponges.
TheHygiena BAX System PCR Assay for E. coli O157:H7 MP is a multiplexed assay that produces clear yes-or-no results for standard and new varieties of E. coli O157:H7 strains in about 3.5 hours of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for raw ground beef, beef trim, spinach, and lettuce sample types. Validated by AFNOR Certification for raw beef, pork, lamb, and chicken; raw milk; fruits and vegetables, and ready-to-eat meals.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Coli produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.