Hooked Weight Set
Supplier: Troemner
30390785
10754-350EA 477.84 USD
10754-350
Hooked Weight Set
Weights Calibration Weights
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Set of nine
  • Variety of sizes
  • Durable plastic case
Units in are in Newtons.
Delivery information: This set includes one 10N unit, one 5N unit, two 2N units, one 1N unit, one 0.5N, two 0.2N units, and one 0.1N unit for a set of nine. It also includes a durable plastic case.