SpeedStreaks™ Loops and Needles, Calibrated, Hardy Diagnostics
Supplier: Hardy Diagnostics
HSND
10753-964EA 172.36 USD
10753-964
SpeedStreaks™ Loops and Needles, Calibrated, Hardy Diagnostics
Spreaders Inoculating Loops and Needles
SpeedStreaksTM are disposable inoculating loops, available in several styles and color coded for convenience.
The 1 μL and 10 μL loops come in two different styles (rigid or flexible) to fit a variety of streaking preferences.
Ordering information: Packaged in Ziploc® bags with certification of calibration.