Petri Dish, No Stacking Ring, 'Slipable', Mono Plate, Sterile
Supplier: Hardy Diagnostics
3301
10753-948EA 123.47 USD
10753-948
Petri Dish, No Stacking Ring, 'Slipable', Mono Plate, Sterile
Culture Dishes Petri Dishes
Petri dish, with smooth bottom, without stacking ring.
  • Manufactured from pure virgin polystyrene
  • Unsurpassed in optical clarity, permitting unimpaired observation of dish content - ideal for micro photography
  • Engineered for optimum flatness to provide uniform agar thickness - reducing media costs
  • Designed with nesting lugs for optimum culture growth and media shelf life
  • Produced in a wide variety of shapes and sizes
Applications: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, pharmaceutical manufacturing
Packaging: Petri dishes are packaged 500 per case.