Microbore Extention Set, Braun
Supplier: B Braun
V5450
10753-942CS 310.81 USD
10753-942
Microbore Extention Set, Braun
Cannulae
Microbore extension set with proximal and distal male luer locks.
  • Removable slide clamp
  • 1.016 mm (0.04") I.D. x 79 cm (31") Length
  • Latex-free