Analytical and Precision Balances, MS-TS
Supplier: Mettler Toledo
30133526
10753-558EA 5195.88 USD
10753-558 10753-560 10753-562 10753-564 10753-566 10753-568 10753-570 10753-698 10753-700 10753-702 10753-704 10753-706 10753-708 10753-710 10753-712 76307-400 76307-402 76307-404 76307-406 76307-408 76307-410 76307-412 76307-414 76307-416 76307-418 76307-420 76307-422 76307-424 76307-426 76307-428
Analytical and Precision Balances, MS-TS
Balances and Scales
METTLER TOLEDO® MS-TS series balances are reliable and robust balances designed for everyday weighing.
  • Reliable and robust
  • Easy to operate
  • Advanced security features
  • Metal housing
  • Easy to clean
Easy to operate units incorporate advanced security features, and these balances make even sophisticated weighing simple and worry free.
To ensure trusted results, the MS-TS series balances feature built-in checks and safety features. All units include MinWeigh warnings, LevelControl, ISO-Log (adjustment history), sample IDs, passcode protection, and Automatic Internal Adjustment (FACT). These easy to use units are supplied standard with a 17.8 cm (7") extra-large color TFT touchscreen, an intuitive user interface, and more than ten built-in applications. The robust and durable design features full metal housing, a robust Monobloc weighing cell, and massive overload protection. Units are easy to clean and chemical-resistant, as the full metal housing is resistant to most chemicals including acetone. A QuickLock feature for draft shields and glass panels allows dismantling in seconds.
Capacities range from 120 to 12200 g, with readabilities starting from 0.1 mg.
Certifications: UL listed and CSA certified.
Ordering information: The two-place readability models do not include draft shields. QuickLock draft shields are only supplied with models featuring three-place readability or better. Optional Service packages are offered to assure continual reliability and maintain optimum balance performance. Proper installation and initial calibration are key to ensuring the balance performs properly from day one. A METTLER TOLEDO® technician performs installation qualification and operational qualification (IQ/OQ). Contact your representative for Service package details.