HardyDisk™ AST Azithromcin, AZM-15, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8051
10753-480EA 35.56 USD
10753-480
HardyDisk™ AST Azithromcin, AZM-15, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ ASTs are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST), also known as disk diffusion or Kirby-Bauer testing.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.