Mycoplasma Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G102
10753-452PK
116.4
USD
10753-452
Mycoplasma Agar, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Mycoplasma media is recommended for use in isolation of mycoplasma species associated with bovine mastitis.
- Optically clear polystyrene - For distortion free viewing
- Heavy-Duty- Reduces breakage during shipping
- Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
Polystyrene Petri plates feature raised rings for stacking that prevent sliding.
Colonies have a "fried egg" appearance when viewed at 100X under the microscope.
Ordering information: Breathable Packaging - Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)