HDL Cholesterol E, Wako
Supplier: Wako Life Sciences
The HDL Cholesterol E kit determines HDL Cholesterol in serum.
- Utilizes a manual precipitation method
- Determines HDL Cholesterol in serum
- 90 tests
The HDL Cholesterol E kit contains reagents utilizing a manual precipitation method for the determination of HDL Cholesterol in serum. Contains 90 tests.