Anti-C16orf5 Rabbit Polyclonal Antibody

Supplier: Prosci
5047_S
10750-436EA 191.02 USD
10750-436
Anti-C16orf5 Rabbit Polyclonal Antibody
Antibodies
CDIP Antibody: The p53 tumor-suppressor gene integrates numerous signals that control cell life and death; loss of its functions contributes to the development of most cancers. CDIP is a novel pro-apoptotic target gene whose inhibition abrogates p53-mediated apoptotic responses. Overexpression of CDIP induced apoptosis in transfected cells while siRNA suppression of caspase-8 mRNA blocked this CDIP-induced apoptosis, indicating that the CDIP-dependent apoptosis pathway proceeds through extrinsic cell death pathway. CDIP may thus represent a novel target for drug design to maximize p53 response and sensitize tumor cells to cancer therapy. Multiple isoforms of CDIP are known to exist.

Type: Primary
Antigen: CDIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR