ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco Products
Supplier: Thermco
ACC064CFC ACC007CFC ACC007FFC ACC008CFC ACC008FFC ACC012CFC ACC012FFC ACC013CFC ACC015CFC ACC015FFC ACC016CFC ACC016FFC ACC017CFC ACC017FFC ACC018CFC ACC018FFC ACC019CFC ACC019FFC ACC020CFC ACC020FFC ACC021CFC ACC022CFC ACC022FFC ACC026CFC ACC028CFC ACC028FFC ACC029CFC ACC030FFC ACC044CFC ACC044FFC ACC045CFC ACC045FFC ACC046CFC ACC046FFC ACC047CFC ACC047FFC ACC056CFC ACC056FFC ACC058CFC ACC058FFC ACC059CFC ACC059FFC ACC060CFC ACC060FFC ACC062CFC ACC062FFC ACC063CFC ACC063FFC ACC064CFC ACC064FFC ACC065CFC ACC065FFC ACC066CFC ACC066FFC ACC067CFC ACC067FFC ACC048CFC ACC048FFC ACC050FFC ACC051FFC ACC054CFC ACC054FFC
10143-072EA 527.67 USD
10143-072 10747-414 10747-416 10747-418 10747-420 10747-434 10747-436 10747-438 10747-444 10747-446 10747-448 10747-450 10747-452 10747-454 10747-456 10747-458 10747-460 10747-462 10747-464 10747-466 10747-468 10747-470 10747-472 10747-474 10747-482 10747-486 10747-488 10747-490 10747-492 10747-494 10747-528 10747-530 10747-532 10747-534 10747-536 10747-538 10747-540 10747-542 10747-544 10747-546 10747-552 10747-554 10747-556 10747-558 10747-560 10747-562 10747-568 10747-570 10747-572 10747-574 10747-576 10747-578 10747-580 10747-582 10747-584 10747-586 10747-588 10747-590 10747-832 10747-834 10747-838 10747-840 10747-842 10747-846 10747-848
ISO17025 ASTM Certified Liquid-in-Glass Mercury Thermometers, Total Immersion, Nos. 7 to 67, Thermco Products
Thermometers Analog Thermometers
These high precision liquid-in-glass mercury thermometers are hand blown, which bear graduations, numbers and letters etched into the glass and filled with and inert gas above the mercury column
ASTM thermometers are used for a variety of specified tests which quantify and qualify repeatable test results in the fields of oil & petroleum testing.
These thermometers were designed for specific testing and can be calibrated to your specific requirements.
Certifications: A NIST Traceable Certificate of Calibration from Thermco Products ISO/IEC accredited laboratory at the specified certification points.
Ordering information: Accessories not included.
Caution: WARNING—Mercury has been designated by EPA and many state agencies as a hazardous material that can cause central nervous system, kidney and liver damage. Mercury, or its vapor, may be hazardous to health and corrosive to materials. Caution should be taken when handling mercury and mercury containing products. See the applicable product Safety Data Sheet (SDS) for details.