Easy 50R Ultrasonic Sieve Cleaner
Supplier: Elma
1120118-001
10724-488EA 1642.52 USD
10724-488
Easy 50R Ultrasonic Sieve Cleaner
Ultrasonic Cleaners
The powerful Elmasonic Easy 50R ultrasonic cleaner has 3 operating modes optimized for cleaning of sieves and lab instruments, degassing of solvents, and preparation of laboratory samples.
  • Ultrasonic bath for all standard laboratory applications
  • 3 optimized modes: Sieve cleaning, Degassing, Sample preparation
  • Intuitive control panel
  • High intensity ultrasonic power
  • Cylindrical tank designed for 8-inch sieves and 1-liter solvent bottles
Dependable, state-of-the-art Elma ultrasonic cleaners have a timer which can be set from 1-30 minutes or continuous operation. For safety purposes the unit turns off automatically after 12 hours of continuous operation. The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run.
Certifications: CE, UL, CSA, ISO9001, ISO13485.
Ordering information: The cover and basket are available and are ordered separately.