Rodent Injection Cone, Plas-Labs™
Supplier: Plas-Labs
561-RC
10718-028EA 661.74 USD
10718-028
Rodent Injection Cone, Plas-Labs™
Animal Supplies Animal Restraints
A unique conical design allows quick and efficient positioning of tailed rodents for repeated sampling
Chemically bonded acrylic construction may be chemically sterilized or washed at a maximum temperature of 82.2°C (180°F).
Warning: Do not autoclave or sterilize with acetone.
The free-standing clear acrylic cone has a tail channel for quick placement of animals from 15g mice to 600g rats. Suction feet secure the rectangular base for benchtop stability. All working edges are smooth finished.