Autoclavable Water Bottle, Tecniplast®
Supplier: Tecniplast Usa
ACBT0150
10717-475EA 35.13 USD
10717-475
Autoclavable Water Bottle, Tecniplast®
Bottles Plastic Bottles
Shatterproof, transparent, autoclavable water bottle with permanent graduations.
Bottles fit under filter-style covers. Designed for cage sizes A and B.