SputEm™ Sputum Collection System, Simport Scientific
Supplier: Simport Scientific
C200G
10713-890CS 429.69 USD
10713-890
SputEm™ Sputum Collection System, Simport Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Sputum Sample Collection
The SputEm™ Sputum Collection System offers an ideal method to collect, carry, and process biological samples
Available in a light green color, three narrow vertical windows allow the contents of the tube to be discreetly viewed. A wide base ensures great stability and prevents tipping. The large collection funnel is made in such a way that specimens fall directly into the graduated centrifuge tube and do not contaminate the outside threads. The centrifuge tube screw cap is kept sterile at all times under the snap cap on top of the funnel.
A patient label is already affixed on top of the unit and can easily be transferred to the tube before leaving for the laboratory. The base can be used as a support during transit.
The unit features a base which incorporates a removable, sterile, 50mL graduated polypropylene conical tube that can withstand centrifugation up to 5000rpm or 3000g.
Certifications: For IVD use. CE certified.
Ordering information: Each unit is sterile and individualy wrapped.