Dawn® Professional Power Dissolver
Supplier: Janitorial Supplies
PGC56037
10713-806CS 207.65 USD
10713-806
Dawn® Professional Power Dissolver
Cleaning Agents Detergents and Surface Cleansers
Professional Power Dissolver makes the dishes virtually spotless with a liquid spray grease dissolver.
  • For washing dishes by hand
  • Liquid spray grease dissolver for dishes
  • No scrubbing required
  • Ideal for use on baked and burned on grease
  • Pretreat your toughest pots and pans and wipe the grease away
  • Effective for pots, pans, dishes, stoves, ovens and grills
No scrubbing required and ideal for use on baked and burned on grease. For use on ceramic, glass, metal, plastic.
It has a chemical specification of Dipropylene Glycol Butoxy Ether, Phenoxyethanol, 2-Aminoethanol, (1-Methylethyl)-, Sodium Salt (1:1), Amine Oxides, C10-16-Alkyldimethyl, Limonene and Benzenesulfonic Acid