Anti-EMD Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12982
10711-976EA 590.77 USD
10711-976
Anti-EMD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of EMD.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: EMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR