Anti-Canine oral papillomavirus E7 protein Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16117
10708-804EA 653.85 USD
10708-804
Anti-Canine oral papillomavirus E7 protein Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Canine oral papillomavirus E7 protein.

Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Canine oral papillomavirus E7 protein
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR