Moeller's Decarboxylase Base
Supplier: HARDY DIAGNOSTICS
Y41
10693-094PK 200.43 USD
10693-094
Moeller's Decarboxylase Base
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of gram negative enteric bacilli.
For the differentiation of gram negative enteric bacilli based on the decarboxylation reaction of amino acids. A control tube for use with Y42, Y43, and Y44.