Male Luer-Lok Priming Syringes, Thermo Scientific
Supplier: Thermo Scientific
365KL531
10662-058EA 150.5 USD
10662-058
Male Luer-Lok Priming Syringes, Thermo Scientific
Syringes Syringes for Chromatography
Male Luer-Lok priming syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings