Sodium dihydrogen phosphate, anhydrous 98.0-102%, Hi-AR™

Supplier: HiMedia

GRM3964-500G
10662-018EA 83.46 USD
10662-018
Sodium dihydrogen phosphate, anhydrous 98.0-102%, Hi-AR™
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2

Order Now

Specification Test Results

Appearance White to off-white crystals or powder or chunks
Solubility 100 mg soluble in 1 mL of water
pH (1M in water at 25°C) 4.00 - 6.00
Chloride (Cl) ≤ 0.014%
Sulfate (SO₄) ≤ 0.15%
Loss on drying (at 60°C for 1h, then 105°C for 4 h) ≤ 2.00%
Assay (NaOH Titration) 98.00 - 102.00%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR