Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
1100
10611-616CS 450.93 USD
10611-616
Optical Lens Cleaner, FoxWest Sales
Cleaning Agents Lens Cleaning Solutions
Optical lens cleaner is a gentle cleaning solution that can be used to clean all optical glass surfaces including microscope optics, absorption cells, and lenses.
This cleaner contains no abrasives or silicones so it will not etch, scratch, or streak.