Silicone Heat Transfer Fluid 510, FoxWest Sales
Supplier: Decon Labs
2101
10611-548EA 1376.22 USD
10611-548
Silicone Heat Transfer Fluid 510, FoxWest Sales
Circulator Bath Fluids
Silicon Heat Transfer Fluid 510 is a methyl-phenyl polysiloxane fluid designed for applications that require ultra-violet light protection or applications that require thermal stability at high temperatures
The working temperature range for silicone bath fluid is 0 to 88°C (32 to 190°F). The fluid viscosity is 14cs.