SP Bel-Art Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18983-0000
10600-116EA 133.55 USD
10600-116
SP Bel-Art Petri Dish Incubation Tray, Bel-Art Products, a part of SP
Trays/Dishes/Pots Laboratory Trays/Dishes
The petri dish incubation tray holds 100 mm (4") petri dishes securely on an incubator rack.
  • Incubation tray is an excellent aid for keeping cell cultures organized
  • Steam autoclavable at 121 °C (250 °F) for 15 to 20 minutes
Each tray has 28 holes in the bottom, 19 mm (3/4") in diameter, to allow air passage. Made from 1.6 mm (1/16") thick, white epoxy-coated steel.