SP Bel-Art No-Wire™ Cuvette Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18520-0000
10600-036EA 35.34 USD
10600-036
SP Bel-Art No-Wire™ Cuvette Rack, Bel-Art Products, a part of SP
Racks Cuvette Racks
All-polypropylene rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.
  • Polypropylene construction
  • Will not scratch glass cuvettes
  • Autoclavable
Forty-two 10 mm light path cuvettes fit in this 6 x 7 row layout. Steam autoclavable at 121ºC (250ºF).