Syringes, Polypropylene, B. Braun
Supplier: B Braun
US4617509F-02
10571-342CS 152.6 USD
10571-342
Syringes, Polypropylene, B. Braun
Syringes General-Purpose Syringes
This syringe is designed for consistent performance
  • High visibility – transparent barrel with bold graduations
  • Secure backstop to prevent accidental plunger withdrawal
  • Double seal stopper to prevent medication leakage
  • Large, stable finger grip
It has double-seal stopper and smooth plunger action.