Anti-EMD Mouse Polyclonal Antibody

Supplier: Abnova
H00002010-B01P
10555-872EA 335 USD
10555-872
Anti-EMD Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR