Anti-GIMAP5 Antibody Pair
Supplier: Abnova
H00055340-PW3
10538-678EA 747.69 USD
10538-678
Anti-GIMAP5 Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-GIMAP5 (300 µl); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-GIMAP5 (50 µl)