Anti-GAPDH Antibody Pair
Supplier: Abnova
H00002597-PW2
10538-604EA 747.69 USD
10538-604
Anti-GAPDH Antibody Pair
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-GAPDH (300 µg); 2. Antibody pair for Western Blot: Rabbit Polyclonal Anti-GAPDH (50 µl)