Anti-FBXO6 Mouse Monoclonal Antibody
Supplier: Abnova
H00026270-PW2
10538-296EA 747.69 USD
10538-296
Anti-FBXO6 Mouse Monoclonal Antibody
Antibodies
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-FBXO6 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-FBXO6 (50 ?g)