Manual Injection Valve, Cytiva
Supplier: Cytiva
29-0239-58
10498-050EA 672.77 USD
10498-050
Manual Injection Valve, Cytiva
Valves Injection Valves
Manual injection valve is a 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column.