Sample Valve, Cytiva
Supplier: Cytiva
29-0238-96
10498-042EA 711.92 USD
10498-042
Sample Valve, Cytiva
Valves Injection Valves
This sample valve allows either the buffer or the sample to enter the flow path.
The sample valve enables the direct application of the sample onto the column using a pump.