Screw Lid Kit, Cytiva
Supplier: Cytiva
11-0004-10
10498-012EA 63.06 USD
10498-012
Screw Lid Kit, Cytiva
Closures Bottle Closures
The Screw Lid Kit is a bottle-top attachment for buffer bottles used with AKTA systems.
The polypropylene lid is sized for GL45 glass bottles with a membrane that allows tubing to be passed through. The kit contains one screw lid and one cap membrane. The membrane should not be used when working with organic solvents.