Superloops, 10 and 50 ml, Cytiva
Supplier: Cytiva
18-1113-81 18-1113-82
10497-994EA 1526.56 USD
10497-994 10497-996
Superloops, 10 and 50 ml, Cytiva
Chromatography Supplies Chromatography Spare Parts
Superloop can be used in conjunction with 7-port valves
  • Flexible: inject large or small sample volumes
  • High reproducibility and recovery
  • No gradient dilution with internal movable seal
The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection. They may also be used to inject pulses of a specific buffer solution. For use with aqueous solutions and alcohols.