Beaker-Style Collection Cup with Pour Spout, Therapak®
Supplier: Therapak, LLC
74848
10497-260CS 487.35 USD
10497-260
Beaker-Style Collection Cup with Pour Spout, Therapak®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
This miniature cup offers convenient collection and pour-off into urinalysis tubes or transport vials.
  • Compact design
  • Pour spout for easy transfer
  • Available with temperature strip
The cup is graduated to 90 mL and can be stacked when being stored prior to collection.