Anti-MARCH11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-9345R-HRP
10489-162EA 803.92 USD
10489-162
Anti-MARCH11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
E3 ubiquitin-protein ligase that mediates polyubiquitination of CD4. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. May play a role in ubuquitin-dependent protein sorting in developmenting spermatids.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: MARCH11
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR