Anti-MARCH5 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-9339R-CY5
10489-094EA 803.93 USD
10489-094
Anti-MARCH5 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
MARCH5 is a member of the E3 ubiquitin-protein ligase family. This mitochonrial E3 ubiquitin-protein ligase plays an importnat role in controlling the morphology of the mitochondria. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. MARCH5 has been shown to interact with MTFN2 and ubiquitinated forms of DRP1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCH5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR