AOAC QuEChERS Triphenylphosphate Solution, Restek
Supplier: RESTEK CORP
31964
10488-704EA 50.3 USD
10488-704
AOAC QuEChERS Triphenylphosphate Solution, Restek
Standards Organic Standards
QuEChERS Standards for AOAC Official Method 2007.01
  • Ready to use for generating test mixes, calibration standards, and spiking experiments.
  • Reliable standards produced according to specifications defined in AOAC Official Method 2007.01.
  • Cost-effective QuEChERS standards can be used without dilutions for greater lab efficiency.
Following QuEChERS methods is even quicker and easier when you use Restek method-specific chemical standards for AOAC Official Method 2007.01 (Pesticide Residues in Foods by Acetonitrile Extraction and Partitioning with Magnesium Sulfate). Our suite of AOAC QuEChERS standards includes internal standards mix, triphenylphosphate (TPP) solution, and QC spike mix. Each standard can be used directly without dilutions because they are formulated to the exact concentrations prescribed by AOAC Method 2007.01.
Contains: Triphenylphosphate (115-86-6)