Anti-AGFG1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-8072R-CY7
10476-182EA 803.92 USD
10476-182
Anti-AGFG1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Required for vesicle docking or fusion during acrosome biogenesis (By similarity). May play a role in RNA trafficking or localization. In case of infection by HIV-1, acts as a cofactor for viral Rev and promotes movement of Rev-responsive element-containing RNAs from the nuclear periphery to the cytoplasm. This step is essential for HIV-1 replication.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGFG1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR