Anti-BRF1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-6358R-CY5.5
10435-798EA 803.92 USD
10435-798
Anti-BRF1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
General activator of RNA polymerase which utilizes different TFIIIB complexes at structurally distinct promoters. The isoform 1 is involved in the transcription of tRNA, adenovirus VA1, 7SL and 5S RNA. Isoform 2 is required for transcription of the U6 promoter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR