IG® RNA Cleanup Kit
Supplier: INTACT GENOMICS, INC MS
4005
104090-580PK 360 USD
104090-580
IG® RNA Cleanup Kit
Enzymes
Useful for cleaning up in vitro transcribed RNA from IG® sgRNA synthesis kits.
  • Rapid and simplified workflow with a single wash buffer
  • Efficiently purify RNA ≥25 nt (a simple modification enables purification of RNA ≥15 nt)
  • Option to elute in 20 to 50 µl for more concentrated RNA than the standard 100 µl
  • Standard 70 to 100% RNA recovery
  • Customizable buffer options for difficult reactions
  • Purified RNA is ready for use in a wide variety of downstream applications including: RT-PCR, RNA library prep for NGS, formation of RNP complexes for genome editing, microinjection, RNA labeling, and transfection
Intact Genomics is your resource for simple and fast cleanup of RNAs including sgRNAs. This kit purifies and concentrates up to 50 µg of RNA from enzymatic reactions. We recommend using this kit following sgRNA synthesis via the IG® sgRNA synthesis kits to remove proteins, salts and most unincorporated nucleotides. The Intact Genomics RNA cleanup kit has a simplified workflow, as shown below, and is ideal for cleanup and concentration of RNA after enzymatic treatments such as DNase I, Proteinase K, labeling, capping or in vitro transcription (IVT).
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.