IG® PE pegRNA Synthesis Kits
Supplier: INTACT GENOMICS, INC MS
The IG® PE pegRNA synthesis kit is simple to use, and is scalable to help you reach necessary yields of pegRNA.
The IG® PE pegRNA Synthesis Kit is a perfect choice for a variety of pegRNA synthesis needs
- IG® PE pegRNA Synthesis Kit includes everything but your target oligos (all buffers, enzymes, scaffolds)
- User simply needs to provide the oligos for your target: a pair of sgRNA oligo and pegRTpbs oligo generated using the template design in this manual
- Rapid workflow (less than 1 hour)
- Scalable yield (100 µl reaction)
Combine the Intact Genomics IG® PE pegRNA synthesis kit with IG® PEmax enzyme for a complete set of reagents for in vitro precision genome editing of your target sequence.
Shipping temperature: Dry ice
Certifications:
Manufactured under ISO 13485:2016 Quality Management certified standards.